ContactUsLinkComponent
VWR* 8-Well Centrifuge Tube Rack
VWR® 8-Well Centrifuge Tube Rack
Catalog #: 82024-452
Supplier:  VWR International
VWR* 8-Well Centrifuge Tube Rack
VWR® 8-Well Centrifuge Tube Rack
Catalog #: 82024-452
Supplier:  VWR International
Specifications
  • Cat. no.:
    82024-452
  • W×D×H:
    14 x 7.7 x 8.8 cm (51/2 x 3 x 31/2")
Specifications
About this item
Rack is ideal for holding up to eight 50 mL conical centrifuge tubes and features a slotted bottom for secure handling.
  • Autoclavable
  • Resistant to chemicals
Rack can also accommodate other types and sizes of tubes. Tubes sit in 30mm square holes. Epoxy-coated steel wire rack is chemical-resistant and autoclavable.
Customers Who Bought This Also Bought