Non-Sterile Spec-Tec™ Stretch Glove Liners, Wells Lamont
Catalog #: 80091-146
Supplier: Wells Lamont
Product available on GSA Advantage®
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:White
- Glove size:S
- Cat. no.:80091-146
- Supplier No.:M114S
Specifications
About this item
The high performance fiber of these glove liners are blended with Lycra® for stretch and reusability.
- Good tactile sensitivity
- Ambidextrous
- High performance fiber and lycra blend
The ambidextrous gloves can be washed up to 12 times without shrinking.
Latex free, the gloves have good tactile sensitivity.
Certifications: ANSI Cut Level 2.
Customers Who Bought This Also Bought