SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
Catalog #: 80080-508
Supplier: SP Industries
Specifications
- Color:Orange
- Material:EVA (Ethylene vinyl acetate)
- Cat. no.:80080-508
- For Tube O.D.:5.0 mm
- Supplier No.:521-ORG-1000
Specifications
About this item
Manufactured from high quality polyethylene or ethylene vinyl acetate to ensure a tight seal.
- Different colors help to track samples
- Available in large quantities for high throughput laboratories
Caution: Please avoid using disposable NMR tube caps when CDCl₃ serves as the reference solution as the material(s) could be dissolved. For CDCl₃, we recommend PTFE tube caps.
Customers Who Bought This Also Bought