Black Phenolic Rubber-Lined Screw Caps, WHEATON®
Catalog #: 80076-402
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:22-400
- Cat. no.:80076-402
- Supplier No.:W239299
Specifications
About this item
Solid-top caps with white, styrene-butadiene rubber liners.
These black phenolic rubber-lined screw caps are made of phenolic resin.
Customers Who Bought This Also Bought