Peptide Vessels, Jacketed with Lower Drain Valves, Chemglass
Catalog #: 80071-408
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:1000 ml
- Description:Reaction vessel, 100 mm (I.D.)
- Height:185 mm
- Cat. no.:80071-408
- Flange Size:100 mm
- Supplier No.:CG-1868-04
Specifications
About this item
Cylindrical, jacketed, peptide synthesis reaction vessel with coarse porosity fritted resin support.
- The vessel is constructed with a flange having a groove formed in the sealing surface of the flange
- Flange is also ground to permit a greased seal with any of the corresponding lids
- Lower outlet is a CG-511 bottom drain valve
Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing.
Ordering information: Supplied complete with two polypropylene hose connections and two GL-14 screw caps (for the clamp see CG-141; for the o-ring see CG-147). Lid not included.
Customers Who Bought This Also Bought