Replacement PPN Hose Connection Set, Chemglass
Catalog #: 80061-452
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Silicone Seals
- Cat. no.:80061-452
- Supplier No.:CG-1563-02
Specifications
About this item
Replacement PPN hose connection set for CG-1560 and CG-1562 DURAN® filter flasks.
Set consists of one each: PPN tubulation (A), Silicone rubber seal (B), Male PPN hose connection (C), Female PPN connection (D).
Packaging: Replacement silicone seals are packaged 10/pack.
Customers Who Bought This Also Bought