DURAN® Filtering Flask, with Replaceable Hose Connection, Chemglass
Catalog #: 80061-434
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:15 L
- Description:Heavy wall flask, with replaceable hose connection
- Cat. no.:80061-434
- Supplier No.:CG-1560-08
Specifications
About this item
Duran® heavy wall filtering flask with a unique removable PPN hose connection.
- Hose connection is secured to the flask via a silicone rubber seal
- Bottle shaped filter flask, uncoated or plastic coated with removable PPN hose connection
BodyTextAccessories: Replacement PPN hose connection set contains PPN tubulation, Silicone rubber seal, Male PPN hose connection and Female PPN connection.
Customers Who Bought This Also Bought