TLC Sprayers and Components, Chemglass
Catalog #: 80060-590
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:TLC Sprayer, Complete Assembly
- Cat. no.:80060-590
- Capacity:125 ml
- Supplier No.:CG-1180-03
Specifications
About this item
These TLC sprayers provide a fine, uniform spray ideally adjusted for the development of chromatographic patterns
Spray pattern is adjustable by varying the coverage of the vent hole with the user’s thumb. Sprayer is used with low pressure air (5/PSI) or rubber pressure bulb. Sprayer consists of an atomizer top attached to the reservoir with a Rodaviss® 24/40 joint. This design provides for a positive, grease-free seal between the atomizer and reservoir and eliminates the glass hooks used on other sprayers. Complete sprayer consists of the atomizer top, Erlenmeyer shaped reservoir, open-top screw cap and #212 Viton o-ring.
Customers Who Bought This Also Bought