Order Entry
ContactUsLinkComponent
BRAND* Semi-Micro Spectrophotometer Cuvettes
BRAND® UV-Cuvette Disposable Spectrophotometer/Photometer Cuvettes, BrandTech
Catalog #: 77776-745
Supplier:  Brandtech
BRAND* Semi-Micro Spectrophotometer Cuvettes
BRAND® UV-Cuvette Disposable Spectrophotometer/Photometer Cuvettes, BrandTech
Catalog #: 77776-745
Supplier:  Brandtech

Specifications

  • Description:
    Semi-Micro Cuvettes
  • Cat. no.:
    77776-745
  • Pathlength:
    10 mm
  • Volume Range:
    1.5–3.0 mL
  • Supplier No.:
    759165

Specifications

About this item

These disposable UV-transparent cuvettes fit most commercial 10mm-pathlength spectrophotometers and photometers. The cuvettes are manufactured from a proprietary plastic that is transparent from 220 - 900 nm, replacing quartz cuvettes for DNA, RNA, protein purity and quantification, and other UV/visible applications, while eliminating scratching, washing and sample cross-contamination.

  • Replaces quartz for many common assays
  • Enhanced chemical compatibility
  • 70 µl minimum fill volume
  • Available individually wrapped

These cuvettes also offer superior chemical compatibility for polar organic solvents, acids, and bases, including acetone, butanone, DMF, and concentrated hydrochloric acid. Cuvettes are mold-cavity matched for the lowest variation in extinction coefficient.

Ultra-micro cuvettes are available in two z-dimensions (8.5mm and 15mm) to match the beam height of most commercial spectrophotometers

The polypropylene cuvette rack features 16 numbered positions and can be autoclaved to 121 °C (250 °F).

A 70 µl minimum fill volume conserves sample. Round caps are available for ultra-micro cuvettes to provide a plug seal for short-term sample storage. Individually wrapped BIO-CERT* ultra-micro cuvettes are DNA-, DNase-, and RNase-free, providing a purity assurance not available with quartz cuvettes.

Customers Who Bought This Also Bought