Order Entry
ContactUsLinkComponent
Sterile Disposable Flask
Nalgene® Disposable Erlenmeyer Flasks with Vented Closure, PETG, Sterile, Baffle Bottom, Thermo Scientific
Catalog #: 74910-018
Sterile Disposable Flask
Nalgene® Disposable Erlenmeyer Flasks with Vented Closure, PETG, Sterile, Baffle Bottom, Thermo Scientific
Catalog #: 74910-018

Specifications

  • Capacity:
    125 ml
  • Description:
    Baffled Bottom
  • Cat. no.:
    74910-018
  • Thread:
    38 - 430
  • Supplier No.:
    4116-0125

Specifications

About this item

Made of lightweight, clear PETG plastic, these flasks are ideal for shaker and suspension cell culture, as well as media preparation or storage.

  • Flasks feature molded-in graduations
  • Non-pyrogenic, non-cytotoxic
  • Sterilized to SAL 10-6

The high-density polyethylene closure has a hydrophobic 0.2µm PTFE membrane that allows sterile air exchange. Sterile disposable flasks reduce the chance of cross contamination.

Customers Who Bought This Also Bought