Order Entry
ContactUsLinkComponent
STIRRER BLENDER 1L2SPD CAN CSA
Two-Speed Lab Blenders, Waring
Catalog #: 74790-002
Supplier:  Waring Products
Two-Speed Lab Blenders, Waring
Catalog #: 74790-002
Supplier:  Waring Products

Specifications

  • Description:
    Two-Speed Laboratory Blender, with Glass Container
  • Electrical:
    120 V
  • Speed Range:
    18000 - 22000 rpm
  • Capacity:
    1 L
  • Supplier No.:
    7011GC

Specifications

About this item

This versatile laboratory blender is applicable for most lab work, providing no-load speeds of 18,000rpm and 22,000rpm

Available with either a heat-resistant glass, or stainless steel 1L container. Both have a handle and two-piece vinyl and styrene lid. Motor housing is epoxy-coated. Rated Amps: 3.6; 120V, 50/60Hz; 0.40HP. UL listed. Also available in heavy-duty versions.

Customers Who Bought This Also Bought