Order Entry
ContactUsLinkComponent
VWR* Hand-Held pen style pH10 measurement instrument
VWR® pH/Temperature Meter, Pen Style
Catalog #: 74730-030
Supplier:  VWR International
VWR* Hand-Held pen style pH10 measurement instrument
VWR® pH/Temperature Meter, Pen Style
Catalog #: 74730-030
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    pH10A Meter
  • Temperature accuracy:
    ±0.3 (±0.6°F)
  • Temperature resolution:
    0.1 °C (0.2 °F) °C
  • Temperature range:
    0.0 to 99.9°C (32.0 to 211.8°F)
  • Display:
    98 × 64 graphic LCD
  • Weight:
    105 g (3.7 oz.)
  • pH accuracy:
    ±0.02 pH ± 1 LSD, ±0.1 (within 10°C of calibration), ±0.2 (within 20°C of calibration)
  • pH range:
    0.00 to 14.00
  • pH resolution:
    0.01
  • Cat. no.:
    74730-030

Specifications

About this item

The model pH10A meter is the perfect instrument for economical spot sampling of pH and temperature in applications such as wastewater, surface water, aquaculture, hydroponics, pools, and education.

  • Automatic calibration and buffer recognition
  • 50-set memory (pH, temp, date and time stamp)
  • Replaceable single- or double-junction electrode
  • GLP functionality
  • 1-, 2-, or 3-point calibration
  • Hold feature locks stable readings on the display
  • >200 hour battery life with low battery indicator

This ultra-compact instrument even includes a graphic display with on-screen instructions. Available with single- or dual-junction electrodes (comes with single-junction electrode). No need to throw this pen-style pH instrument away when the sensor goes bad like some other products. Simply replace the electrode within minutes and keep testing.

Certifications: Waterproof IP-67. CE compliance.
Ordering information: Instrument includes model 606110 pH electrode, batteries, and instruction manual. Manufacturer's one-year warranty.
Customers Who Bought This Also Bought