Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Baffle Bottom, Thermo Scientific
Catalog #: 73520-404
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:500 ml
- Cat. no.:73520-404
- Thread:45 - 430
- Supplier No.:4113-0500
Specifications
About this item
Graduated flasks are made of light, crystal-clear PETG plastic with easy-to-open polyethylene screw closures.
- Disposing of flasks reduces chance of cross-contamination
- Non-pyrogenic, non-cytotoxic
- Sterilized to SAL 10-6
The leak-proof high-density polyethylene screw closures open to vent with 1/4 turn.
Ideal for shaker and suspension cell culture, media preparation, or storage
Packaging: Individually packaged for easy storage.
Customers Who Bought This Also Bought