Nalgene® Serum Vial Closures, HDPE, Thermo Scientific
Catalog #: 73520-094
Supplier: Thermo Fisher Scientific
Specifications
- Sterility:Sterile
- Color:Green
- Description:Lined Closures
- Cat. no.:73520-094
- Supplier No.:342158-0024
Specifications
About this item
These 20mm, high-density polyethylene screw cap closures are available lined or unlined and sterile or nonsterile.
Ordering information: All closures are bulk packed. Sterile closures are bulk packed in a case with a three-ply liner. Unlined closures require a stopper (not included).
Customers Who Bought This Also Bought