Nalgene® Diagnostic Bottles, PETG, Sterile, Thermo Scientific
Catalog #: 73520-078
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:5 ml (0.17 oz.)
- Cat. no.:73520-078
- Packaging:Tray-Packed
- Supplier No.:2035-0005
Specifications
About this item
PETG single-use diagnostic bottles are ideal for sampling, storage, and shipment of reagents and buffer solutions.
- Bottles and caps are gamma sterilized to 10⁻⁶ sterility assurance levels and have a lot number
- Nonpyrogenic, noncytotoxic
Ordering information: Supplied with white, lined screw caps with a 20-415 thread finish. Tray-packed bottles are packed 20 bottles per tray, 5 trays per case. Bulk-packed bottles are packed 100 bottles per tray, 5 trays per case. All trays are packaged in double-lined cartons for cleanroom use, with caps assembled.
Customers Who Bought This Also Bought