Order Entry
ContactUsLinkComponent
Nalgene High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Catalog #: 73520-072
Nalgene High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Nalgene® High-Density Polyethylene Bottles, Sterile, Narrow Mouth, Thermo Scientific
Catalog #: 73520-072

Specifications

  • Cap Size:
    24-415
  • Capacity:
    250 ml (8.5 oz.)
  • Cat. no.:
    73520-072
  • Supplier No.:
    342089-0008

Specifications

About this item

HDPE bottles feature a narrow mouth and leakproof closure design.

Manufactured and packaged in controlled environments to minimize biological and particulate contamination. Gamma irradiated to 10⁻⁶ sterility assurance levels.

Certifications: Bottles meet USP-VI and are noncytotoxic and nonpyrogenic.
Ordering information: Supplied with white polypropylene closures.
Customers Who Bought This Also Bought