Order Entry
ContactUsLinkComponent
2-Tube/Slide Foam LabMailer*
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Catalog #: 73320-066
Supplier:  Sonoco Thermosafe
2-Tube/Slide Foam LabMailer*
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Catalog #: 73320-066
Supplier:  Sonoco Thermosafe

Specifications

  • Description:
    2 tube specimen mailer, 107×16 mm
  • Cat. no.:
    73320-066
  • Tube Dimensions:
    107×16 mm
  • Dimensions:
    10.7×1.6 cm (47/32×5/8")
  • Supplier No.:
    362

Specifications

About this item

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials during specimen transport.

For further protection, optional mailing sleeves, specifically designed to fit foam mailers, are available. Sleeves are made of sturdy, 18-point, white paperboard and feature reverse tuck closures that enable the foam mailer to withstand multiple trips. Sleeves may be custom-printed with lab name, address, and postal permit number.

  • For VacuTainer®, Microtainer®, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam saves postage
  • Reusable and recyclable

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials during specimen transport.

A unique array of specimen shippers fills the need for protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.

Customers Who Bought This Also Bought