Order Entry
ContactUsLinkComponent
DRY ICE CHEST 2.5CU FT ORD W/
Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Catalog #: 73320-006
Supplier:  Sonoco Thermosafe
DRY ICE CHEST 2.5CU FT ORD W/
Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Catalog #: 73320-006
Supplier:  Sonoco Thermosafe

Specifications

  • Description:
    Hard-cased Dry Ice Chest without Handles
  • Wall Thickness:
    21/4"
  • Interior Dimensions:
    17L x 161/2W x 163/8H"
  • Exterior Dimensions:
    23L x 22W x 203/4H"
  • Capacity:
    2.7 cu.ft.
  • Supplier No.:
    303

Specifications

About this item

ThermoSafe Storage Chests provide mobile, non-electrical, low temperature maintenance for a wide variety of laboratory and field applications.

  • Deluxe containers provide years of trouble-free service
  • Rugged polyethylene interior and exterior casing with super-efficient, non-deteriorating urethane foam insulation
  • Rounded interior corners for easy clean-up
  • Spring-loaded handles fold down when not in use

These dry ice chests are ideal for demanding cryogenic and laboratory testing, as well as the transport and storage of temperature sensitive specimens.

Customers Who Bought This Also Bought