Order Entry
ContactUsLinkComponent
Disposable Plastic Fracture and Safety Collar
Ampule Crackers, Dyn-A-Med Products
Catalog #: 72960-034
Supplier:  Dyn-A-Med
Ampule Crackers, Dyn-A-Med Products
Catalog #: 72960-034
Supplier:  Dyn-A-Med

Specifications

  • Description:
    Ampule Cracker
  • Cat. no.:
    72960-034
  • For:
    1 - 4 ml ampules
  • Supplier No.:
    81014

Specifications

About this item

Designed to facilitate opening a pre-scored glass ampule.

  • Disposable
  • Non Sterile
  • Plastic fracture and safety collar

Available in two versions: For 1 - 4 ml ampules and for 5 - 10 ml ampules.

Customers Who Bought This Also Bought