Automation Pipette Tips for Qiagen BioRobot Workstations, Thermo Scientific
Catalog #: 72830-336
Supplier: Thermo Scientific
Specifications
- Volume:1100 µl
- Packed:96 Tips/Tray, 50 Trays/Case
- Filter:+
- Tip Style:Filtered
- For Use With:Qiagen BioRobot automated workstations
- Supplier No.:953-253
Specifications
About this item
Thermo Scientific provides a solution for your Qiagen BopRobot pipette tips.
These automation pipette tips have been designed, validated, and tested on a Qiagen BioRobot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our BioRobot workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Customers Who Bought This Also Bought