BioRobotix® Pipette Tips for Tecan® and Cavro® Workstations, Molecular BioProducts
Catalog #: 72830-132
Supplier: Thermo Scientific
Specifications
- Volume:20 µl
- Packed:96/Tray, 50 trays
- Tip Style:Liquid Sensing
- Supplier No.:901-253
Specifications
About this item
Compatible with Tecan®, Cavro®, and Packard MultiPROBE II HT style workstations.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Total of six volumes available, for both tip styles, for increased repeatability in any application. Filter tips utilize the ART® barrier for increased protection against contamination.
Customers Who Bought This Also Bought