Closures for Storage/Reaction Vials with Closure, Chemglass
Catalog #: 71000-556
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:SURE-LINK™ Cap, Open Top, 24-414, polypropylene, green, TFE septa
- Cat. no.:71000-556
- Supplier No.:CG-4911-20