Chlorine Test Papers, Lamotte Chemical Products
Catalog # 66170-326
Supplier: LaMotte
Small Business Enterprise
Specifications
- Description:Chlorine Test Paper
- Cat. no.:66170-326
- Range:10, 50, 100, 200 ppm
- Supplier No.:4250-BJ
- No. of Tests:200 test papers
Specifications
About this item
Chlorine test papers are chemically treated to indicate sanitizer levels.
Frequently Bought Together