Order Entry
ContactUsLinkComponent
DPD No.4R Tablet
DPD Chlorine Comparator Kit and Test Tablets, LaMotte
Catalog #: 66130-379
Supplier:  LaMotte
Small Business Enterprise
CAS Number:  
DPD No.4R Tablet
DPD Chlorine Comparator Kit and Test Tablets, LaMotte
Catalog #: 66130-379
Supplier:  LaMotte
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    Chlorine DPD #4, Reagent Tablets
  • Cat. no.:
    66130-379
  • Supplier No.:
    6906A-H

Specifications

About this item

Comparator test kit employs diethyl-p-phenylene diamine (DPD) indicator in convenient tablet form to measure the various factors of water chlorination

The DPD No. 1R tablet measures free chlorine, the DPD No. 3R tablet shows dichloramine content and total chlorine level, and the DPD No. 4R tablet contains reagents needed for one-step reading of total residual chlorine. Drop a single tablet in the test sample and compare the resulting color reaction against the permanent color standards in the comparator. Color standard values are 0.2, 0.4, 0.6, 0.8, 1.0, 1.5, 2.0 and 3.0ppm chlorine.

Ordering information: Kit is supplied with comparator, two 10mL glass test tubes, EPA-approved manual No. 330.5, and 50 tablets each of No. 1R and No. 3R. Additional tablets sold separately.
Customers Who Bought This Also Bought