Screw Caps for Screw-Thread Sample Storage Vials, Thermo Scientific
Catalog #: 66030-775
Supplier: Thermo Scientific
Specifications
- Description:Screw Caps, 18-400, PTFE/PE Foam Liner
- Material:PP
- Cap Color:White
- Supplier No.:B7815-18