Order Entry
ContactUsLinkComponent
Scintillation Vial Rack
WHEATON® Vial Racks, DWK Life Sciences
Catalog #: 66022-525
Small Business Enterprise
Scintillation Vial Rack
WHEATON® Vial Racks, DWK Life Sciences
Catalog #: 66022-525
Small Business Enterprise

Specifications

  • Dimensions:
    33L x 17W x 3H cm (12.9 x 6.7 x 1.2")
  • Description:
    50-Position Rack, with 28.1 mm Well I.D.
  • No. of wells:
    5 x 10
  • Cat. no.:
    66022-525
  • Supplier No.:
    868806

Specifications

About this item

Manufactured from polypropylene, vial racks are easy to clean and autoclavable

Each rack features alphanumeric indexing to simplify identification and selection. The racks are sturdy and stackable, even when vials are in place.

Ordering information: When selecting the proper rack, the vial O.D. cannot exceed the well I.D. dimension.
Customers Who Bought This Also Bought