White Plastic Screw Caps, WHEATON®
Catalog #: 66021-840
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:22-400
- Cap Type:Polypropylene
- Cat. no.:66021-840
- Liner Material:Foil
- Supplier No.:240804
Specifications
About this item
White plastic screw caps fit Wheaton vials (see 66021-679 series) and other glass scintillation vials.
- Suitable for marking
- Not autoclavable
- Available unlined or choose from a variety of liners
Customers Who Bought This Also Bought