Order Entry
ContactUsLinkComponent
PTFE/Silicone Rubber Septa
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Catalog #: 66010-774
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Catalog #: 66010-774

Specifications

  • Ø:
    13.5 mm
  • Description:
    PTFE-faced silicone rubber septa
  • Thickness:
    0.005" PTFE, 0.060" silicone rubber
  • Cat. no.:
    66010-774
  • Supplier No.:
    73818-15

Specifications

About this item

Ideal for use with open-top caps to access container contents with a syringe.

  • Resists coring
  • Highly chemically-resistant PTFE facing maintains integrity of content
  • Silicone rubber backing allows repeated puncturing through the seal
  • Recommended for use in all autosamplers using screw thread vials
Customers Who Bought This Also Bought