Polypropylene Screw Caps, Linerless, Kimble Chase
Catalog #: 66010-680
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:15-415
- Color:Natural
- Supplier No.:73805B-15415
Specifications
About this item
Polypropylene caps feature one-piece construction with a seal achieved by a unique interior design.
Caps withstand repeated autoclaving when loosened or removed from tubes.
Certifications: Designed from ASTM Specification E982
Customers Who Bought This Also Bought