Phenolic Screw Thread Closure, Kimble Chase
Catalog #: 66010-672
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:15-425
- Color:Black
- Height:—
- Cat. no.:66010-672
- Supplier No.:73804 15425
Specifications
About this item
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought