Order Entry
ContactUsLinkComponent
Cryogenic Vial (Non-Printed, Bulk Pack), PP, non sterile
Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Catalog #: 66008-748
Cryogenic Vial (Non-Printed, Bulk Pack), PP, non sterile
Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Catalog #: 66008-748

Specifications

  • Cap included:
    Yes
  • Capacity:
    2.00 ml
  • Cat. no.:
    66008-748
  • Diameter ×H:
    13.5×38 mm
  • Supplier No.:
    5011-0020

Specifications

About this item

Nalgene cryogenic vials are externally threaded vial for aseptic technique, using a design similiar to the linerless closures used on Nalgene bottles and carboys.

  • Self-standing and may be used without a holder

They are an economical, non-sterile version of Cat. Nos. 5000-0012 and -0020 without printing or graduations.

Ordering information: Closures and vials are packed separately.
Caution: For research use only; not for in-vitro diagnosis or parenterals. Do not autoclave.
Customers Who Bought This Also Bought