Order Entry
ContactUsLinkComponent
System 100 Cryogenic Vial, PP, sterile
Nalgene® SYSTEM 100™ Cryogenic Vials, Thermo Scientific
Catalog #: 66008-710
System 100 Cryogenic Vial, PP, sterile
Nalgene® SYSTEM 100™ Cryogenic Vials, Thermo Scientific
Catalog #: 66008-710

Specifications

  • Cap included:
    Yes
  • Capacity:
    1.50 ml
  • Diameter ×H:
    12×48 mm
  • Supplier No.:
    5000-1020

Specifications

About this item

These vials are designed to increase storage capacity from 81 to 100 vials in mechanical and vapor-phase (LN₂) freezers when used with SYSTEM 100 CryoBox™ vial boxes.

  • Externally threaded vials for aseptic technique
  • White marking area, graduations, and fill line printed on vials
  • Radiation sterilized
  • Non-cytotoxic and non-pyrogenic
  • Self-standing

They are constructed of polypropylene and can withstand temperatures from –196 to 121 °C (–321 to 250 °F). Silicone gasketed polypropylene closures ensure leakproof performance in a microcentrifuge (up to 8000 g) and during shipment.

Certifications: CE marked.
Packaging: Packaged 25 vials per bag, 20 bags per case.
Customers Who Bought This Also Bought