Versilon™ C-219-A Flexible Chemical Transfer Tubing
Catalog #: 63009-348
Supplier: Saint Gobain Life Sciences
Specifications
- Tubing I.D.:⅛"
- Tubing O.D.:¼"
- Wall thickness:¹⁄₁₆"
- Durometer hardness:Shore A, 67
- Supplier No.:ABH02007
Specifications
About this item
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Customers Who Bought This Also Bought