VWR® Latex Rubber Tubing, Black
Catalog #: 62995-608
Supplier: VWR International
Specifications
- Tubing I.D.:⅛"
- Tubing O.D.:³⁄₁₆"
- Wall thickness:1/32"
- Description:50' Reel
- Cat. no.:62995-608
Specifications
About this item
Black latex rubber tubing withstands repeated sterilization and has superior tensile strength.
- Durometer hardness: Shore A, 35
- Tensile strength: 3500 psi
- Packaged on 50' reels or in box packs
Black rubber tubing has maximum resistance to aging. Withstands repeated sterilization.
Customers Who Bought This Also Bought