Order Entry
ContactUsLinkComponent
VWR* Red Vacuum Rubber Tubing
VWR® Red Vacuum Rubber Tubing
Catalog #: 62995-059
Supplier:  VWR International
VWR* Red Vacuum Rubber Tubing
VWR® Red Vacuum Rubber Tubing
Catalog #: 62995-059
Supplier:  VWR International

Specifications

  • Wall thickness:
    1/8"
  • Cat. no.:
    62995-059
  • Coil Length:
    50 ft.
  • I.D.×O.D.:
    ¼×½"

Specifications

About this item

Natural rubber tubing with extra-heavy wall especially recommended for vacuum work.

  • Durometer hardness: Shore A, 50
  • Tensile strength: 2000 to 2600 psi

Has high resilience and good tensile and tear strength.

Customers Who Bought This Also Bought