Humidity/Temperature Monitor, Sper Scientific
Catalog #: 61220-512
Supplier: Sper Scientific
Specifications
- Description:Humidity/Temperature Monitor with Certification
- Cat. no.:61220-512
- Supplier No.:800016C
Specifications
About this item
Digital humidity temperature monitor with a large display.
- Available with or without certification
Simultaneously monitors both temperature and humidity on an extra large 3-3/16 x 2-1/2" dual display which can be read from across a room. Monitor humidity and temperature in labs, chemical storage areas, clean rooms, stock rooms, warehouses, factories, greenhouses, and outdoors. Recall min/max temperature and min/max humidity readings over any time period or have it automatically reset itself every 24 hours. Stand or wall mount.
Certifications: N.I.S.T. traceable certificate of compliance.
Ordering information: Comes with 1 AAA battery which lasts a full year.
Customers Who Bought This Also Bought