VWR® Traceable® Jumbo-Display Dial Thermometers
Catalog #: 61161-310
Supplier: VWR International
Specifications
- Probe:133 × 3.2 mm (51/4 × 1/8")
- Description:Digital Dial Thermometer
- Temperature resolution:0.1° from –20 to 200°, 1° outside this range
- Temperature range:–50 to 150°C (–58 to 302°F)
- Accuracy:±1 °C between –50 and 150 °C
- Length:16.5 cm (61/2")
Specifications
About this item
Thermometers fit cuvettes, test tubes, flasks, and beakers and will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids.
Thermometers fit cuvettes, test tubes, flasks, and beakers. Will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids. Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4cm (21/8") diameter and features a large, bright 1cm (3/8") high LCD.
- Traceable®
- Wide temperature range
- Easy-to-read LCD
- Read °C and °F
Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4 cm (21/8") diameter and features a large, bright 1 cm (3/8") high LCD.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with replaceable 1.5V silver-oxide battery.
Customers Who Bought This Also Bought