VWR® Films for ELISA and General Incubation, Polyester
Catalog #: 60941-120
Supplier: VWR International
Specifications
- Optical property:Transparent
- Sterility:Non sterile
- Description:Sealing films, polyester
- Thickness:77,5 µm
Specifications
About this item
These 77.5 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and well-to-well contamination, and eliminating 'edge effects' in sensitive ELISA assays.
- Functional temperature range from −40 to +120 °C
- Available sterile and non sterile
- Non pierceable
Polyester films can also be used with cell culture plates for short-term storage, incubation, and containment of biohazards.
Each film L×D: 146,1×79,4 mm
Customers Who Bought This Also Bought