VWR® Plate Roller
Catalog #: 60941-118
Supplier: VWR International
Specifications
- Description:VWR® Plate Roller
- Cat. no.:60941-118
Specifications
About this item
This 10.2 cm (4") soft rubber roller is designed for assuring tight and consistent application of adhesive films and foils to microplates.
- Tight and consistent application
- Secure seal around all microplate wells
Customers Who Bought This Also Bought