PYREX® Flat Bottom Culture Tubes, Rimless, Corning
Catalog #: 60856-130
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:39 ml
- Ø:25 mm
- Environmentally Preferable:Low Manufacturing Impact
- Height:100 mm
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Supplier No.:9850-25
Specifications
About this item
Flat-bottom culture tubes are ideal for plant tissue and general culture work.
Two-position polypropylene closures are sold separately. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
Flat-bottom culture tubes are ideal for plant tissue and general culture work.
Two-position polypropylene closures are sold separately. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage.
Ordering information: Tubes and closures sold separately.
Customers Who Bought This Also Bought