Corning® Reusable Phenolic Caps, Corning
Catalog #: 60827-271
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Cap Size:24-410
- Color:Black
- Description:Reusable Phenolic Cap with Rubber Liner
- Environmentally Preferable:Low Manufacturing Impact
- Liner Material:Rubber
- Supplier No.:9999-241
- Environmental certification:I-RECs, ISO 14001 - Monterrey
Specifications
About this item
These reusable Corning® caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving.
Caps have GPI threads and are provided with a glued-in white rubber liner.
Customers Who Bought This Also Bought