Order Entry
ContactUsLinkComponent
PYREX* Culture Tube
PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Catalog #: 60826-836
Supplier:  Corning
Product available on GSA Advantage®
PYREX* Culture Tube
PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Catalog #: 60826-836
Supplier:  Corning
Product available on GSA Advantage®

Specifications

  • Capacity:
    6 ml
  • Ø:
    12 mm
  • Description:
    Round-bottom, rimless culture tube
  • Height:
    75 mm
  • Supplier No.:
    99445-12

Specifications

About this item

Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.

These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed.

Customers Who Bought This Also Bought