VWR® Dual-Position Caps, PE
Catalog #: 60819-091
Supplier: VWR International
Specifications
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
- Cat. no.:60819-091
- For Tubes:17 x 100 mm
Specifications
About this item
White caps for use with VWR® and most popular brand disposable culture tubes.
- Polyethylene fabrication
- Dual-position caps
- Compatible with most disposable culture tubes
Caps permit aspiration of cultures.
Customers Who Bought This Also Bought