Order Entry
ContactUsLinkComponent
SGE GC Inlet Liner
SGE Inlert Liners, GC, Trajan Scientific and Medical
Catalog #: 60362-506
SGE GC Inlet Liner
SGE Inlert Liners, GC, Trajan Scientific and Medical
Catalog #: 60362-506

Specifications

  • O.D.:
    6.3 mm
  • Description:
    Split/Splitless, GC Inlet Liner, FocusLiner, 78.5 mm (4 mm I.D.)
  • Cat. no.:
    60362-506
  • Supplier No.:
    092002

Specifications

About this item

This wide range of fully deactivated inlet liners is compatibile with the industry's most popular instrument makes and models.

Note: Varian® 1177 injectors accept all Agilent Technologies 6.3mm O.D. liners.

Liners cover a variety of applications, and ensure complete sample transfer from syringe to column without loss. The improved designs result in increased precision and accuracy of sample transfer.

Varian® 1177 injectors accept all Agilent Technologies 6.3 mm O.D. liners.

Customers Who Bought This Also Bought