Synthware Septa, Teflon®-Lined Silicone Rubber
Catalog #: 60005-060
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Ø:8 mm
- For:Side port hickman still head
- Material:Teflon®-lined silicone rubber
- Supplier No.:R1408
Specifications
About this item
Replacement PTFE lined silicone septums.
- Silicone construction
- Ptfe lined
For use with open top closures.
Customers Who Bought This Also Bought