[ST] Joint Clips
Catalog #: 60004-954
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Plastic joint clip
- ST Joint Size:34/45
- Supplier No.:CS003445
Specifications
About this item
Made of plastic.
- Heavy duty plastic construction
- Color coded
- Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Customers Who Bought This Also Bought