Synthware PTFE Stoppers, Solid and Hollow
Catalog #: 60004-902
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint No.:14/20
- Supplier No.:S531420
Specifications
About this item
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.
Customers Who Bought This Also Bought