Synthware Round-Bottom Flasks, Single-Neck, Kemtech America
Catalog #: 60002-298
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Capacity:100 ml
- ST Joint No.:14/20
- Cat. no.:60002-298
- Supplier No.:F301100
Specifications
About this item
Heavy wall design.
- Heavy wall borosilicate construction
- Ground glass joint
- Various sizes available
Hand-blown from tubing to ensure uniform wall thickness.
Customers Who Bought This Also Bought