Synthware Distillation Connecting Adapters
Catalog #: 60000-596
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60000-596
- Bottom [ST] Joint Size:14/20
- Top ST Joint Size:[ST] 14/20
- Supplier No.:A351420
Specifications
About this item
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
- Inner joints at bottom and side arm
- Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Customers Who Bought This Also Bought