Synthware Adapters, Vacuum or Argon, Kemtech America
Catalog #: 60000-166
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:24/40
- Description:Adapter
- Cat. no.:60000-166
- Supplier No.:A292440
Specifications
About this item
Use this adapter for vacuum or argon applications.
- Inner [ST] joint
- Side hose connection
Featuring a hose connection on the side and inner [ST] joint. Hose ext. Ø is 8 mm.
Customers Who Bought This Also Bought